missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-amino-2-methoxybenzoate, 98%

Product Code. 11440773
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440773 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11440773 Supplier Thermo Scientific Alfa Aesar Supplier No. A13239.14
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 27492-84-8 | C9H11NO3 | 181.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifica

CAS 27492-84-8
Melting Point 154°C to 157°C
Molecular Formula C9H11NO3
MDL Number MFCD00017202
Quantity 25 g
Synonym methyl 4-amino-o-anisate, 4-amino-2-methoxybenzoic acid methyl ester, benzoic acid, 4-amino-2-methoxy-, methyl ester, methyl 2-methoxy-4-aminobenzoate, methyl 4-amino-2-methoxy-benzoate
InChI Key YUPQMVSYNJQULF-UHFFFAOYSA-N
SMILES COC(=O)C1=CC=C(N)C=C1OC
IUPAC Name methyl 4-amino-2-methoxybenzoate
Molecular Weight (g/mol) 181.19
PubChem CID 168705
Formula Weight 181.19
Percent Purity 98%
Chemical Name or Material Methyl 4-amino-2-methoxybenzoate
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • Methyl 4-amino-2-methoxybenzoate
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Ergänzende Informationen
  • MIXTURE LIST-Contain: Methyl 4-amino-o-anisate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.