missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-aminosalicylate, 97%, Thermo Scientific Chemicals

Product Code. 15412337
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15412337 100 g 100g
15422337 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15412337 Supplier Thermo Scientific Alfa Aesar Supplier No. 045520.22
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

Methyl 4-aminosalicylate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4136-97-4
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00088091
InChI Key QQOXBFUTRLDXDP-UHFFFAOYSA-N
Synonym methyl 4-aminosalicylate, methyl p-aminosalicylate, pas methyl ester, p-aminosalicylic acid methyl ester, methyl 4-amino-2-hydroxybenzenecarboxylate, benzoic acid, 4-amino-2-hydroxy-, methyl ester, methyl 4-amino-2-hydroxy-benzoate, salicylic acid, amino-, methyl ester, 4-aminosalicylic acid methyl ester, 4-amino-2-hydroxy-benzoic acid methyl ester
PubChem CID 77787
ChEBI CHEBI:35089
IUPAC Name methyl 4-amino-2-hydroxybenzoate
SMILES COC(=O)C1=C(C=C(C=C1)N)O

Specifications

Quantity 100 g
Solubility Information Slightly soluble in water.
Formula Weight 167.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Methyl 4-aminosalicylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.