missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(bromomethyl)benzoate, 98%

Product Code. 13498508 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13498508 100 g Glass bottle 100g
10743541 5 g Glass bottle 5g
10771925 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13498508 Supplier Thermo Scientific Acros Supplier No. 306101000
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2417-72-3 | C9H9BrO2 | 229.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2417-72-3
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00032453
InChI Key NLWBJPPMPLPZIE-UHFFFAOYSA-N
PubChem CID 256687
IUPAC Name methyl 4-(bromomethyl)benzoate
SMILES COC(=O)C1=CC=C(CBr)C=C1

Specifications

Melting Point 53.0°C to 58.0°C
Color White to Yellow
Boiling Point 130.0°C to 135.0°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2C6H4CO2CH3
Quantity 100 g
Synonym methyl 4-bromomethyl benzoate, 4-bromomethylbenzoic acid methyl ester, methyl4-bromomethyl benzoate, benzoic acid, 4-bromomethyl-, methyl ester, 4-bromomethyl benzoic acid methyl ester, p-methoxycarbonyl benzyl bromide, methyl p-bromomethyl benzoate, 4-bromomethyl-benzoic acid methyl ester, 4-carbomethoxybenzyl bromide
Formula Weight 229.07
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Methyl 4-(bromomethyl)benzoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.