missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(bromomethyl)benzoate, 98%

Product Code. p-3758350 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10743541

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2417-72-3 | C9H9BrO2 | 229.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2417-72-3
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00032453
InChI Key NLWBJPPMPLPZIE-UHFFFAOYSA-N
PubChem CID 256687
IUPAC Name methyl 4-(bromomethyl)benzoate
SMILES COC(=O)C1=CC=C(CBr)C=C1

Specifications

Melting Point 53.0°C to 58.0°C
Color White to Yellow
Boiling Point 130.0°C to 135.0°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2C6H4CO2CH3
Quantity 5 g
Synonym methyl 4-bromomethyl benzoate, 4-bromomethylbenzoic acid methyl ester, methyl4-bromomethyl benzoate, benzoic acid, 4-bromomethyl-, methyl ester, 4-bromomethyl benzoic acid methyl ester, p-methoxycarbonyl benzyl bromide, methyl p-bromomethyl benzoate, 4-bromomethyl-benzoic acid methyl ester, 4-carbomethoxybenzyl bromide
Formula Weight 229.07
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Methyl 4-(bromomethyl)benzoate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.