missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-chloro-3-nitrobenzoate, 98%

Product Code. 11381897
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381897 5 g 5g
11391897 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381897 Supplier Thermo Scientific Alfa Aesar Supplier No. L07407.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14719-83-6 | C8H6ClNO4 | 215.59 g/mol

Methyl 4-chloro-3-nitrobenzoate, is used as starting material for the preparation of the methyl benzoates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-chloro-3-nitrobenzoate, is used as starting material for the preparation of the methyl benzoates

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14719-83-6
Molecular Formula C8H6ClNO4
Molecular Weight (g/mol) 215.59
MDL Number MFCD00016989
InChI Key XRTKWPWDSUNLHS-UHFFFAOYSA-N
Synonym methyl 4-chloro-3-nitrobenzate, 4-chloro-3-nitrobenzoic acid methyl ester, methyl-4-chloro-3-nitrobenzoate, benzoic acid, 4-chloro-3-nitro-, methyl ester, acmc-209cya, methyl-3-nitro-4-chlorobenzoate, methyl 3-nitro-4-chlorobenzoate, xrtkwpwdsunlhs-uhfffaoysa, methyl 4-chloro-3-nitro-benzoate, methyl4-chloro-3-nitrobenzoate
PubChem CID 735797
IUPAC Name methyl 4-chloro-3-nitrobenzoate
SMILES COC(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O

Specifications

Melting Point 78°C to 82°C
Quantity 5 g
Beilstein 1643729
Solubility Information Soluble in water.
Formula Weight 215.59
Percent Purity 98%
Chemical Name or Material Methyl 4-chloro-3-nitrobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.