missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-fluoro-2-methoxybenzoate, 97+%, Thermo Scientific Chemicals

Product Code. p-7034677
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15439057

Brand: Thermo Scientific Alfa Aesar H52370.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Methyl 4-fluoro-2-methoxybenzoate  is used as a catalyst, kinase inhibitors and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-fluoro-2-methoxybenzoate  is used as a catalyst, kinase inhibitors and pharmaceutical intermediates.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 204707-42-6
Molecular Formula C9H9FO3
Molecular Weight (g/mol) 184.17
MDL Number MFCD06203990
InChI Key LJUAEPNTXDJBRX-UHFFFAOYSA-N
Synonym 4-fluoro-2-methoxybenzoic acid methyl ester, methyl 2-methoxy-4-fluoro-benzoate, benzoic acid, 4-fluoro-2-methoxy-, methyl ester, rarechem al bf 1023, methyl-4-fluoro-2-methoxybenzoate, methyl 4-fluoranyl-2-methoxy-benzoate, 2-methoxy-4-fluorobenzoic acid methyl ester, 4-fluoro-2-methoxy-benzoic acid methyl ester
PubChem CID 10241478
IUPAC Name methyl 4-fluoro-2-methoxybenzoate
SMILES COC(=O)C1=C(OC)C=C(F)C=C1

Specifications

Quantity 250 mg
Formula Weight 184.17
Percent Purity ≥97%
Chemical Name or Material Methyl 4-fluoro-2-methoxybenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.