missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-hydroxy-3-iodobenzoate, 98%
Beskrivelse
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tekniske data
Tekniske data
| CAS | 15126-06-4 |
| Melting Point | 155°C to 159°C |
| Color | Brown to White |
| Molecular Formula | C8H7IO3 |
| MDL Number | MFCD05664378 |
| Quantity | 5 g |
| Sensitivity | Light sensitive |
| Synonym | methyl4-hydroxy-3-iodobenzoate, methyl 3-iodo-4-hydroxybenzoate, methyl 4-hydroxy-3-iodo-benzoate, 4-hydroxy-3-iodobenzoic acid methyl ester, 4-hydroxy-3-iodo-benzoic acid methyl ester, benzoic acid, 4-hydroxy-3-iodo-, methyl ester, 4-hydroxy-3-iodo-benzoicacidmethylester, acmc-209zk0, ksc495e2f, methyl 4-hydroxy-3-iodbenzoate |
| InChI Key | PXNOLLHARLSLHY-UHFFFAOYSA-N |
| SMILES | COC(=O)C1=CC(=C(C=C1)O)I |
| Vis mere |
RUO – Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?