missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-hydroxy-3-nitrobenzoate, 98+%

Product Code. 11454233
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454233 50 g 50g
11464233 250 g 250g
11444233 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454233 Supplier Thermo Scientific Alfa Aesar Supplier No. A10880.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 99-42-3 | C8H7NO5 | 197.146 g/mol

4-alkoxy-3-nitrobenzoic acids were obtained by the alkylation of methyl 4-hydroxy-3-nitrobenzoate with corresponding n-alkylbromides in 3-pentanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-alkoxy-3-nitrobenzoic acids were obtained by the alkylation of methyl 4-hydroxy-3-nitrobenzoate with corresponding n-alkylbromides in 3-pentanone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-42-3
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00014695
InChI Key GNCWCTBHZCBXGL-UHFFFAOYSA-N
Synonym methyl 3-nitro-4-hydroxybenzoate, benzoic acid, 4-hydroxy-3-nitro-, methyl ester, 4-hydroxy-3-nitrobenzoic acid methyl ester, methyl 3-nitro-4-hydroxy toluate, aurora 6897, 3-nitro-4-hydroxybenzoic acid methyl ester, methyl-4-hydroxy-3-nitrobenzoic acid, 3-nitro-4-hydroxymethylbenzoate, acmc-209scn, 3-10-00-00378 beilstein handbook reference
PubChem CID 66835
IUPAC Name methyl 4-hydroxy-3-nitrobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)O)[N+](=O)[O-]

Specifications

Melting Point 73°C to 77°C
Quantity 50 g
Beilstein 2115197
Formula Weight 197.15
Percent Purity ≥98%
Chemical Name or Material Methyl 4-hydroxy-3-nitrobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.