missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-hydroxybenzoate, 99%

Product Code. 13418348 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle
Unit Size:
100g
10kg
2.5kg
500g
This item is not returnable. View return policy

Product Code. 13418348

Brand: Thermo Scientific Acros 126960100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-76-3 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 99-76-3
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002352
InChI Key LXCFILQKKLGQFO-UHFFFAOYSA-N
Synonym methylparaben, methyl paraben, methyl p-hydroxybenzoate, nipagin, methyl parahydroxybenzoate, maseptol, p-hydroxybenzoic acid methyl ester, 4-hydroxybenzoic acid methyl ester, p-carbomethoxyphenol, tegosept m
PubChem CID 7456
ChEBI CHEBI:31835
IUPAC Name methyl 4-hydroxybenzoate
SMILES COC(=O)C1=CC=C(C=C1)O

Especificaciones

Melting Point 125.0°C to 128.0°C
Color White
Boiling Point 298.6°C
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic Drum
Linear Formula HOC6H4CO2CH3
Quantity 10 kg
Beilstein 10,158
Merck Index 15,618
Solubility Information Solubility in water: 0.25% (25°C). Other solubilities: freely soluble in alcohol,acetone and ether,slightly soluble in benzene and ccl4
Formula Weight 152.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-hydroxybenzoate, 99%
Mostrar más Mostrar menos
compliance-icons
Palabra de advertencia
  • Warning
Clasificación
  • LONG-TERM AQUATIC HAZARD Chronic 2
Indicaciones de peligro
  • H411-Toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado