missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Hydroxybenzoate (Methyl Paraben), TRC

Product Code. 30137123
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137123 100 g 100g
30132964 10 g 10g
17920615 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137123 Supplier Toronto Research Chemicals Supplier No. TRCM325660100G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-76-3
Molecular Formula C8 H8 O3
Molecular Weight (g/mol) 152.15
Synonym Methyl 4-hydroxybenzoate, Methyl parahydroxybenzoate, Nifuroxazide Imp. B (EP), Ethyl Parahydroxybenzoate Imp. B (EP), Butyl Parahydroxybenzoate Imp. B (EP), Propyl Parahydroxybenzoate Imp. B (EP), Methyl Paraben, Methylparaben, Benzoic acid, 4-hydroxy-, methyl ester, Benzoic acid, p-hydroxy-, methyl ester (6CI,8CI), 4-(Carbomethoxy)phenol, 4-(Methoxycarbonyl)phenol, 4-Hydroxybenzoic acid methyl ester, 4-Hydroxymethyl benzoate, Danisol M, E 218, E 218 (preservative), Killitol, Maseptol, Mekkings M, Metaben, Metagin, Methaben, Methyl 4-(3'-butenyloxy)benzoate, Methyl Butex, Methyl Parasept, Methyl chemosept, Methyl p-hydroxybenzoate, Methylben, Metoxyde, Moldex, NSC 3827, NSC 406127, Nipagin, Nipagin M, Para M, Paraben M, Paridol, Preserval, Preserval M, Septos, Solbrol, Solbrol M, Tegasept Methyl, Tegosept M, p-Carbomethoxyphenol, p-Hydroxybenzoic acid methyl ester, p-Methoxycarbonylphenol
IUPAC Name methyl 4-hydroxybenzoate
SMILES COC(=O)c1ccc(O)cc1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C8H8O3/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5,9H,1H3
Formula Weight 152.0473
Percent Purity >95%
Chemical Name or Material Methyl Parahydroxybenzoate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.