missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Hydroxybenzoate (Methyl Paraben), TRC

Product Code. 17920615
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17920615 1 g 1g
30132964 10 g 10g
30137123 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17920615 Supplier Toronto Research Chemicals Supplier No. TRCM3256601G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-76-3
InChI Formula InChI=1S/C8H8O3/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5,9H,1H3
IUPAC Name methyl 4-hydroxybenzoate
Molecular Formula C8 H8 O3
Molecular Weight (g/mol) 152.15
Chemical Name or Material Methyl Parahydroxybenzoate

Specifications

Quantity 1 g
Synonym Methyl 4-hydroxybenzoate, Methyl parahydroxybenzoate, Nifuroxazide Imp. B (EP), Ethyl Parahydroxybenzoate Imp. B (EP), Butyl Parahydroxybenzoate Imp. B (EP), Propyl Parahydroxybenzoate Imp. B (EP), Methyl Paraben, Methylparaben, Benzoic acid, 4-hydroxy-, methyl ester, Benzoic acid, p-hydroxy-, methyl ester (6CI,8CI), 4-(Carbomethoxy)phenol, 4-(Methoxycarbonyl)phenol, 4-Hydroxybenzoic acid methyl ester, 4-Hydroxymethyl benzoate, Danisol M, E 218, E 218 (preservative), Killitol, Maseptol, Mekkings M, Metaben, Metagin, Methaben, Methyl 4-(3'-butenyloxy)benzoate, Methyl Butex, Methyl Parasept, Methyl chemosept, Methyl p-hydroxybenzoate, Methylben, Metoxyde, Moldex, NSC 3827, NSC 406127, Nipagin, Nipagin M, Para M, Paraben M, Paridol, Preserval, Preserval M, Septos, Solbrol, Solbrol M, Tegasept Methyl, Tegosept M, p-Carbomethoxyphenol, p-Hydroxybenzoic acid methyl ester, p-Methoxycarbonylphenol
SMILES COC(=O)c1ccc(O)cc1
Formula Weight 152.0473
Percent Purity >95%
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.