missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-hydroxybenzoate-ring-13C6 solution, 10 μg/mL in acetone, Honeywell Fluka™

Product Code. 15921161 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15921161 5 mL 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15921161 Supplier Honeywell Fluka Supplier No. R13475ML
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

10 μg/mL in acetone. Analytical standard, reference material

Ensure you have the information you need to quickly start your analysis, and pass relevant audits:

  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
CAS 1581694-95-2, 67-64-1
Density 0.790 - 0.792 g/cm3
Flash Point -17°C
Physical Form Liquid
Boiling Point 55°C to 57°C
Industry Type Parabenes
Quantity 5 mL
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.