missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-hydroxybenzoate, sodium salt, 99%, Thermo Scientific Chemicals

Product Code. 10338370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10338370 500 g Plastic bottle 500g
10053673 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10338370 Supplier Thermo Scientific Acros Supplier No. 235145000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5026-62-0 | Molecular Formula: C8H7NaO3 | Molecular Weight: 174.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5026-62-0
Molecular Formula C8H7NaO3
Molecular Weight (g/mol) 174.13
MDL Number MFCD00016470
InChI Key PESXGULMKCKJCC-UHFFFAOYSA-M
Synonym methylparaben sodium, sodium methylparaben, sodium 4-methoxycarbonyl phenolate, preserval ms, methyl 4-hydroxybenzoate, sodium salt, solparol, bonomold omna, methyl 4-hydroxybenzoate sodium salt, sodium 4-carbomethoxyphenolate, unii-cr6k9c2nhk
PubChem CID 23663626
IUPAC Name sodium;4-methoxycarbonylphenolate
SMILES COC(=O)C1=CC=C(C=C1)[O-].[Na+]

Specifications

Melting Point >125°C
Color White
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 500 g
Merck Index 10,5977
Solubility Information Solubility in water: 333g/L (20°C)
Formula Weight 174.13
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Methyl 4-hydroxybenzoate, sodium salt, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 4-hydroxybenzoate, sodium salt
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.