missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-hydroxyphenylacetate, 99%

Product Code. 10762441 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10762441 100 g Glass bottle 100g
10032650 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10762441 Supplier Thermo Scientific Acros Supplier No. 208551000
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14199-15-6 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14199-15-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00002387
InChI Key XGDZEDRBLVIUMX-UHFFFAOYSA-N
Synonym methyl 4-hydroxyphenylacetate, methyl 2-4-hydroxyphenyl acetate, 4-hydroxyphenylacetic acid methyl ester, benzeneacetic acid, 4-hydroxy-, methyl ester, methyl 4-hydroxyphenyl acetate, 4-hydroxyphenylacetic acid methl ester, acetic acid, p-hydroxyphenyl-, methyl ester, p-hydroxyphenylacetic acid methyl ester, 4-hydroxybenzeneacetic acid methyl ester, 4-hydroxy-phenyl-acetic acid methyl ester
PubChem CID 518900
ChEBI CHEBI:68078
IUPAC Name methyl 2-(4-hydroxyphenyl)acetate
SMILES COC(=O)CC1=CC=C(C=C1)O

Specifications

Melting Point 55°C to 58°C
Color White to Yellow
Boiling Point 162°C to 163°C (5.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HOC6H4CH2CO2CH3
Quantity 100 g
Beilstein 10, 191
Formula Weight 166.18
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material Methyl 4-hydroxyphenylacetate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 4-hydroxyphenylacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.