missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-iodobenzoate, 98%

Product Code. 10636794 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10636794 5 g Glass bottle 5g
10085810 25 g Glass bottle 25g
10137420 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10636794 Supplier Thermo Scientific Acros Supplier No. 414460050
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 619-44-3 | C8H7IO2 | 262.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-44-3
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.04
MDL Number MFCD00016353
InChI Key DYUWQWMXZHDZOR-UHFFFAOYSA-N
Synonym methyl p-iodobenzoate, 4-iodobenzoic acid methyl ester, benzoic acid, 4-iodo-, methyl ester, 4-iodo-benzoic acid methyl ester, benzoic acid, p-iodo-, methyl ester, methyl-4-iodobenzoate, methyl 4-iodo-benzoate, methyl-4-iodo-benzoate, pubchem20378, methyl para-iodobenzoate
PubChem CID 69273
IUPAC Name methyl 4-iodobenzoate
SMILES COC(=O)C1=CC=C(C=C1)I

Specifications

Melting Point 113°C to 117°C
Color Beige to Pink
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CO2CH3C6H4I
Quantity 5 g
Formula Weight 262.04
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-iodobenzoate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 4-iodobenzoate
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.