missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Methoxysalicylate, TRC

Product Code. 30469215
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469215 100 g 100g
30474282 25 g 25g
30482149 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30469215 Supplier Toronto Research Chemicals Supplier No. TRCM265515100G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5446-02-6
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.17
Synonym 2-Hydroxy-4-methoxybenzoic Acid Methyl Ester, 2-Hydroxy-p-anisic Acid Methyl Ester, Methyl 2-Hydroxy-4-methoxybenzoate, Methyl 4-Methoxy-2-hydroxybenzoate, NSC 17154
IUPAC Name methyl 2-hydroxy-4-methoxybenzoate
SMILES COC(=O)c1ccc(OC)cc1O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C9H10O4/c1-12-6-3-4-7(8(10)5-6)9(11)13-2/h3-5,10H,1-2H3
Formula Weight 182.0579
Chemical Name or Material Methyl 4-Methoxysalicylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.