missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-methyl-3-nitrobenzoate, 99%, Thermo Scientific™

Product Code. 11420898
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420898 100 g 100g
11400898 5 g 5g
11410898 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420898 Supplier Thermo Scientific Alfa Aesar Supplier No. B25299.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Methyl 4-methyl-3-nitrobenzoate is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7356-11-8
MDL Number MFCD00130054

Specifications

Melting Point 49°C to 53°C
Molecular Formula C23H14O
Quantity 100 g
Synonym 4-Methyl-3-nitrobenzoic acid methyl ester; Methyl 3-nitro-p-toluate
Solubility Information Soluble in methanol.
InChI Key FGCOCPAQFSREBF-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(=O)C2=C3C=CC4=CC=CC5=C4C3=C(C=C5)C=C2
IUPAC Name phenyl(pyren-1-yl)methanone
Molecular Weight (g/mol) 306.36
PubChem CID 13012745
Formula Weight 195.18
Percent Purity 99%
Chemical Name or Material Methyl 4-methyl-3-nitrobenzoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.