missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-methylsalicylate, 98+%

Product Code. p-7056051
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15402159

Brand: Thermo Scientific Alfa Aesar H64793.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4670-56-8 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4670-56-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00020130
InChI Key UITFCFWKYAOJEJ-UHFFFAOYSA-N
Synonym methyl 4-methylsalicylate, methyl-4-methylsalicylate, benzoic acid, 2-hydroxy-4-methyl-, methyl ester, 4-methylsalicylic acid methyl ester, methyl-4-methyl salicylate, methyl m-cresotate, 2-hydroxy-4-methylbenzoic acid methyl ester, methyl 2-hydroxy-4-methyl-benzoate, 2-hydroxy-4-methyl-benzoic acid methyl ester, methyl methylsalicylate
PubChem CID 78400
IUPAC Name methyl 2-hydroxy-4-methylbenzoate
SMILES CC1=CC(=C(C=C1)C(=O)OC)O

Specifications

Melting Point 27°C to 28°C
Color Brown to Yellow
Density 1.147 g/mL
Boiling Point 242°C (literature)
Refractive Index 1.537
Quantity 5 g
Beilstein 2690247
Formula Weight 166.18
Percent Purity ≥98%
Physical Form Low Melting Solid
Chemical Name or Material Methyl 4-methylsalicylate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.