missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-[(Pyridine-3-carbonylamino)methyl]benzoate, TRC

Product Code. 30464468
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30464468 100 mg 100mg
30480429 500 mg 500mg
30477722 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30464468 Supplier Toronto Research Chemicals Supplier No. TRCM643668100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554407-47-5
Molecular Formula C15H14N2O3
Molecular Weight (g/mol) 270.28
SMILES COC(=O)c1ccc(CNC(=O)c2cccnc2)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H14N2O3/c1-20-15(19)12-6-4-11(5-7-12)9-17-14(18)13-3-2-8-16-10-13/h2-8,10H,9H2,1H3,(H,17,18)
Chemical Name or Material Methyl 4-[(Pyridine-3-carbonylamino)methyl]benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.