missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-tert-butylphenylacetate, 96%

Product Code. p-7082135
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469613 5 g 5g
11479613 25 g 25g
11489613 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469613 Supplier Thermo Scientific Alfa Aesar Supplier No. A12421.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3549-23-3 | C13H18O2 | 206.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3549-23-3
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.29
MDL Number MFCD00051913
InChI Key HXVTYMWVMVKVTF-UHFFFAOYSA-N
Synonym methyl p-tert-butylphenylacetate, methyl 4-tert-butylphenylacetate, methyl 2-4-tert-butyl phenyl acetate, 4-tert-butylphenylacetic acid methyl ester, benzeneacetic acid, 4-1,1-dimethylethyl-, methyl ester, unii-8a4h5qo9oj, methyl 2-4-tert-butylphenyl acetate, 8a4h5qo9oj, methyl 4-1,1-dimethylethyl benzeneacetate, acetic acid, p-tert-butylphenyl-, methyl ester
PubChem CID 605629
IUPAC Name methyl 2-(4-tert-butylphenyl)acetate
SMILES COC(=O)CC1=CC=C(C=C1)C(C)(C)C

Specifications

Density 0.994
Boiling Point 149°C to 151°C (30 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.4995
Quantity 5 g
Beilstein 2502880
Formula Weight 206.29
Percent Purity 97%
Chemical Name or Material Methyl 4-tert-butylphenylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.