missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-chloro-2-hydroxybenzoate, 97%, Thermo Scientific™

Product Code. 11448847
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448847 10 g 10g
11438847 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448847 Supplier Thermo Scientific Alfa Aesar Supplier No. A19971.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4068-78-4
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00016461
InChI Key KJWHRMZKJXOWFC-UHFFFAOYSA-N
Synonym methyl 5-chlorosalicylate, benzoic acid, 5-chloro-2-hydroxy-, methyl ester, rarechem al bf 0041, 5-chloro-2-hydeoxybenzoic acid methyl ester, 5-chloro-2-hydroxybenzoic acid methyl ester, acmc-209uvv, methyl-5-chlorosalicylate, methyl 5-chloro-salicylate, methyl-5-chloro-salicylate, 5-chlorosalicylic methyl ester
PubChem CID 77684
IUPAC Name methyl 5-chloro-2-hydroxybenzoate
SMILES COC(=O)C1=C(C=CC(=C1)Cl)O

Specifications

Melting Point 44°C to 48°C
Flash Point >110°C (230°F)
Quantity 10 g
Formula Weight 186.6
Percent Purity 97%
Chemical Name or Material Methyl 5-chloro-2-hydroxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.