missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-fluoro-2-nitrobenzoate, 98%

Product Code. 15467828
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467828 25 g 25g
15457828 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467828 Supplier Thermo Scientific Alfa Aesar Supplier No. H61307.14
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 393-85-1 | C8H6FNO4 | 199.14 g/mol

Methyl 5-fluoro-2-nitrobenzoate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 5-fluoro-2-nitrobenzoate is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water. Soluble in dimethyl sulfoxide and acetone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 393-85-1
Molecular Formula C8H6FNO4
Molecular Weight (g/mol) 199.14
MDL Number MFCD03425619
InChI Key LCAUVFVOLNRVKG-UHFFFAOYSA-N
Synonym 2-nitro-5-fluorobenzoic acid methyl ester, 5-fluoro-2-nitrobenzoic acid methyl ester, 2-nitro-5-fluorobenzoic acid, methyl ester, benzoic acid, 5-fluoro-2-nitro-, methyl ester, methyl 5fluoro-2-nitrobenzoate, methyl5-fluoro-2-nitrobenzoate, 2-nitro-5-fluorobenzoic acid,methyl ester, 3-fluoro-6-nitrobenzoic acid methyl ester, 5-fluoro-2-nitro-benzoic acid methyl ester, benzoic acid,5-fluoro-2-nitro-, methyl ester
PubChem CID 5152049
IUPAC Name methyl 5-fluoro-2-nitrobenzoate
SMILES COC(=O)C1=C(C=CC(F)=C1)[N+]([O-])=O

Specifications

Melting Point 35°C to 37°C
Color White
Quantity 25 g
Solubility Information Slightly soluble in water. Soluble in dimethyl sulfoxide and acetone.
Formula Weight 199.14
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material Methyl 5-fluoro-2-nitrobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.