missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-iodosalicylate, 99%

Product Code. 11499537
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499537 10 g 10g
11409547 50 g 50g
11419547 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499537 Supplier Thermo Scientific Alfa Aesar Supplier No. B22048.09
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 4068-75-1 | C8H7IO3 | 278.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4068-75-1
Molecular Formula C8H7IO3
Molecular Weight (g/mol) 278.045
MDL Number MFCD00016462
InChI Key NRSWJTRJHPRZMH-UHFFFAOYSA-N
PubChem CID 248910
IUPAC Name methyl 2-hydroxy-5-iodobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)I)O

Specifications

Melting Point 75°C to 78°C
Quantity 10 g
Sensitivity Light sensitive
Synonym methyl 5-iodosalicylate, 5-iodosalicylic acid methyl ester, 2-hydroxy-5-iodobenzoic acid methyl ester, benzoic acid, 2-hydroxy-5-iodo-, methyl ester, methyl 2-hydroxy-5-iodobenzenecarboxylate, salicylic acid, 5-iodo-, methyl ester, methyl5-iodosalicylate, methyl-5-iodosalicylate, acmc-209jg0, methyl2-hydroxy-5-iodobenzoate
Formula Weight 278.05
Percent Purity 99%
Chemical Name or Material Methyl 5-iodosalicylate
compliance-icons
Product Identifier
  • Methyl 5-iodosalicylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Methyl 5-iodosalicylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.