missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-(4-Fluorophenyl)-4-isopropyl-2-methylthio-1,4-dihydropyrimidine-5-carboxylate Hydrochloride, TRC

Product Code. 30474782
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474782 25 mg 25mg
30468971 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30474782 Supplier Toronto Research Chemicals Supplier No. TRCM30391025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 6-(4-Fluorophenyl)-4-isopropyl-2-methylthio-1,4-dihydropyrimidine-5-carboxylate Hydrochloride
Molecular Formula C16H19FN2O2S . HCl
Quantity 25 mg
Synonym 6-(4-Fluorophenyl)-1,4-dihydro-4-(1-methylethyl)-2-(methylthio)-5-pyrimidinecarboxylic acid Methyl Ester Hydrochloride
InChI Formula InChI=1S/C16H19FN2O2S.ClH/c1-9(2)13-12(15(20)21-3)14(19-16(18-13)22-4)10-5-7-11(17)8-6-10;/h5-9,13H,1-4H3,(H,18,19);1H
SMILES FC1=CC=C(C2=C(C(OC)=O)C(C(C)C)N=C(SC)N2)C=C1.Cl
IUPAC Name methyl 6-(4-fluorophenyl)-4-isopropyl-2-(methylthio)-1,4-dihydropyrimidine-5-carboxylate hydrochloride
Molecular Weight (g/mol) 358.86
Formula Weight 358.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.