missing translation for 'onlineSavingsMsg'
Learn More

Methyl adipoyl chloride, 96%

Product Code. 11396916
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
Product Code. Quantity unitSize
11396916 2 g 2g
11306926 10 g 10g
11316926 50 g 50g
3 options
This item is not returnable. View return policy

Product Code. 11396916

Brand: Thermo Scientific Alfa Aesar L04377.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35444-44-1 | C7H11ClO3 | 178.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35444-44-1
Molecular Formula C7H11ClO3
Molecular Weight (g/mol) 178.61
MDL Number MFCD00013661
InChI Key HDLGIEZOMYJKAK-UHFFFAOYSA-N
Synonym hexanedioic acid monochloride monomethyl ester, hexanoic acid, 6-chloro-6-oxo-, methyl ester, 6-methoxy-6-oxohexanoyl chloride, methyl 5-chloroformyl pentanoate, 5-chlorocarbonyl-pentanoic acid methyl ester, mono-methyl adipoyl chloride, 5-carbomethoxyvaleryl chloride, methyl 6-chloro-6-oxocaproate, methyl adipoyl chloride
PubChem CID 520737
IUPAC Name methyl 6-chloro-6-oxohexanoate
SMILES COC(=O)CCCCC(Cl)=O

Specifications

Density 1.05
Boiling Point 76°C (0.8 mmHg)
Linear Formula CH3O2C(CH2)4COCl
Refractive Index 1.447
Quantity 2 g
UN Number UN3265
Beilstein 1766187
Sensitivity Moisture sensitive
Formula Weight 178.62
Percent Purity 96%
Chemical Name or Material Methyl adipoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.