missing translation for 'onlineSavingsMsg'
Learn More

Methyl benzoylformate, 99%

Product Code. 10771745 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10771745 5 g Glass bottle 5g
10685172 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10771745 Supplier Thermo Scientific Acros Supplier No. 126370050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 15206-55-0 | C9H8O3 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15206-55-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00008443
InChI Key YLHXLHGIAMFFBU-UHFFFAOYSA-N
Synonym methyl benzoylformate, methyl phenylglyoxylate, methyl oxophenylacetate, methyl phenylglyoxalate, benzoylformic acid methyl ester, methylbenzoylformate, phenylglyoxylic acid methyl ester, phenylglyoxylic acid, methyl ester, benzeneacetic acid, .alpha.-oxo-, methyl ester, unii-23f2045stg
PubChem CID 84835
ChEBI CHEBI:84256
IUPAC Name methyl 2-oxo-2-phenylacetate
SMILES COC(=O)C(=O)C1=CC=CC=C1

Specifications

Melting Point 16°C
Color Yellow
Density 1.1500g/mL
Boiling Point 246°C to 248°C
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5COCO2CH3
Refractive Index 1.5250 to 1.527
Quantity 5 g
Beilstein 10, 657
Specific Gravity 1.15
Solubility Information Solubility in water: 2.02g/L (20°C)
Formula Weight 164.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl benzoylformate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.