missing translation for 'onlineSavingsMsg'
Learn More

Methyl formate, Reagent Grade, 97%, Honeywell™

Product Code. 15667950 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 1 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15667950 1 L Glass bottle 1 L
15677950 2.5 L Glass bottle 2.5 L
15687950 4 x 1 L Glass bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15667950 Supplier Honeywell Research Chemicals Supplier No. M468371L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 107-31-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00003291
InChI Key TZIHFWKZFHZASV-UHFFFAOYSA-N
Synonym formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3
PubChem CID 7865
ChEBI CHEBI:77699
IUPAC Name methyl formate
SMILES COC=O

Tekniske data

Color Colorless
Density 0.974g/mL at 20°C
Boiling Point 34°C (1013 hPa)
Flash Point −19°C (−2.2°F)
Packaging Glass bottle
Linear Formula HCO2CH3
Quantity 1 L
UN Number UN1243
Beilstein 1734623
Formula Weight 60.05g/mol
Percent Purity 0.97
Physical Form Liquid
Chemical Name or Material Methyl formate
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.