missing translation for 'onlineSavingsMsg'
Learn More

Methyl formate, 98%, for spectroscopy

Product Code. 10742453 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10742453 5 mL Glass bottle 5mL
10445851 500 mL Glass bottle 500mL
10414315 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10742453 Proveedor Thermo Scientific Acros N.º de proveedor 414340050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 107-31-3 | C2H4O2 | 60.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

for UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 107-31-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00003291
InChI Key TZIHFWKZFHZASV-UHFFFAOYSA-N
Synonym formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3
PubChem CID 7865
ChEBI CHEBI:77699
IUPAC Name methyl formate
SMILES COC=O

Especificaciones

Acidity 0.0008 meq/g max.
Absorbance 0.009 max. at 320nm, 0.032 max. at 300nm, 0.051 max. at 270nm, 1 max. at 260nm
Melting Point -100.0°C
Density 0.9680g/mL
Boiling Point 31.0°C to 33.0°C
Flash Point −32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HCO2CH3
Refractive Index 1.3420 to 1.3440
Quantity 5 mL
Beilstein 02, 18
Residue after Evaporation 0.0005% max.
Fieser 05,446
Merck Index 15, 6148
Specific Gravity 0.968
Solubility Information Solubility in water: 300g/L (20°C). Other solubilities: soluble in almost all conventional organic solvent,miscible with alcohol,soluble in ether
Viscosity 0.35 mPa.s (20°C)
Formula Weight 60.05
Percent Purity 98%
Grade Spectroscopy
Physical Form Liquid
Chemical Name or Material Methyl formate, 98%
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • Methyl formate
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 1
  • Specific target organ toxicity Category 2
Indicaciones de peligro
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contain: Methyl formate

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.