Learn More
Methyl 2-bromobenzoate, 99%
Description
Methyl 2-bromobenzoate under Heck reaction with acrylonitrile results in lactonization to give the benzofuran-2-one 3-(cyanomethyl)-2-coumaranone. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl 2-bromobenzoate under Heck reaction with acrylonitrile results in lactonization to give the benzofuran-2-one 3-(cyanomethyl)-2-coumaranone. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals
Solubility
Insoluble in water.
Notes
Store in a cool place. Keep away from oxidizing agents.
Specifications
Specifications
| Density | 1.532 |
| Boiling Point | 252°C |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.559 |
| Quantity | 500 g |
| Beilstein | 1862501 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 215.05 |
| Percent Purity | 99% |
| Chemical Name or Material | Methyl 2-bromobenzoate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.