missing translation for 'onlineSavingsMsg'
Learn More

Methyl hydrogen azelate, 96%

Product Code. 11307995
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307995 5 g 5g
11317995 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307995 Supplier Thermo Scientific Alfa Aesar Supplier No. L08116.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2104-19-0 | C10H18O4 | 202.25 g/mol

Methyl hydrogen azelate is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl hydrogen azelate is used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2104-19-0
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00004431
InChI Key VVWPSAPZUZXYCM-UHFFFAOYSA-N
Synonym methyl hydrogen azelate, monomethyl azelate, azelaic acid monomethyl ester, nonanedioic acid, monomethyl ester, monomethyl nonanedioate, 8-carbomethoxyoctanoic acid, azelaic acid, monomethyl ester, mono-methyl azelate, unii-xdi67hf5bh, xdi67hf5bh
PubChem CID 75009
IUPAC Name 9-methoxy-9-oxononanoic acid
SMILES COC(=O)CCCCCCCC(=O)O

Specifications

Melting Point 22°C to 24°C
Boiling Point 146°C to 147°C (1 mmHg)
Flash Point >110°C (230°F)
Linear Formula CH3OOC(CH2)7COOH
Refractive Index 1.447
Quantity 5 g
Beilstein 1775786
Solubility Information Not miscible in water.
Formula Weight 202.25
Percent Purity 96%
Chemical Name or Material Methyl hydrogen azelate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.