missing translation for 'onlineSavingsMsg'
Learn More

Methyl Linolenate, SPEX CertiPrep™

Product Code. 16359557 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16359557 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16359557 Supplier SPEX Certiprep Supplier No. S4222

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Methyl linolenate301-00-81000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging 2mL amber prescored glass ampule
CAS 301-00-8
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Molecular Formula CH4O
MDL Number MFCD00004595
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Synonym methyl linolenate, linolenic acid methyl ester, methyl alpha-linolenate, linolenic acid, methyl ester, unii-0s1ns923k6, alpha-linolenic acid methyl ester, methyl all-cis-9,12,15-octadecatrienoate, methyl 9z,12z,15z-octadeca-9,12,15-trienoate, 9,12,15-octadecatrienoic acid, methyl ester, z,z,z, methyl cis,cis,cis-octadec-9,12,15-trienoate
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
Solvent or Matrix Methanol-P&T
Molecular Weight (g/mol) 32.04
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Methyl linolenate
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.