missing translation for 'onlineSavingsMsg'
Learn More

Methyl methacrylate 1000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30317767
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30317767 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317767 Supplier Dr. Ehrenstorfer Supplier No. DREGA09011080ME
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methacrylic acid-methyl ester
CAS 80-62-6
Analyte or Component Names Methacrylic acid-methyl ester
Shipping Condition Ice pack (-20°C)
Molecular Formula C5 H8 O2
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Basic Butylated Methacrylate Copolymer Imp. B (EP), Methacrylic acid methyl ester (6CI,8CI), Methacrylic acid, methyl ester (7CI), 2-(Methoxycarbonyl)-1-propene, 2-Methyl-2-propenoic acid methyl ester, ACR-PA 20, Acryester M, Acryester MMA, Acryloid HT 100, DL 101, Light Ester M, MMA, Methyl 2-methacrylate, Methyl 2-methyl-2-propenoate, Methyl 2-methylacrylate, Methyl isobutenoate, Methyl methacrylate, NSC 4769, Paraloid HT 100, Pegalan, TEB 3K, Three bond 3057J, Basic Butylated Methacrylate Copolymer Impurity B
InChI Formula InChI=1S/C5H8O2/c1-4(2)5(6)7-3/h1H2,2-3H3
SMILES COC(=O)C(=C)C
IUPAC Name methyl 2-methylprop-2-enoate
Solvent or Matrix Methanol
Molecular Weight (g/mol) 100.12
Quantity 1 mL
Formula Weight 100.0524 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.