missing translation for 'onlineSavingsMsg'
Learn More

Methyl-p -hydroxybenzoate, 99.83%, MP Biomedicals™

Product Code. 11423580 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423580 1 kg 1kg
11433580 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11423580 Supplier MP Biomedicals Supplier No. 0210234101
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 99-76-3
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
InChI Key LXCFILQKKLGQFO-UHFFFAOYSA-N
Synonym methylparaben, methyl paraben, methyl p-hydroxybenzoate, nipagin, methyl parahydroxybenzoate, maseptol, p-hydroxybenzoic acid methyl ester, 4-hydroxybenzoic acid methyl ester, p-carbomethoxyphenol, tegosept m
PubChem CID 7456
ChEBI CHEBI:31835
IUPAC Name methyl 4-hydroxybenzoate
SMILES COC(=O)C1=CC=C(C=C1)O

Specifications

Color White
Quantity 1 kg
Formula Weight 152.1
Percent Purity 99.83%
Physical Form Crystalline Powder
Chemical Name or Material Methyl-p-hydroxybenzoate
compliance-icons compliance-icons
Product Identifier
  • Methyl p-Hydroxybenzoate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Methyl p-Hydroxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.