missing translation for 'onlineSavingsMsg'
Learn More

Methyl-p -hydroxybenzoate, 99.83%, MP Biomedicals™

Product Code. 11433580 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433580 5 kg 5kg
11423580 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11433580 Supplier MP Biomedicals Supplier No. 0210234105
  • ONLINE EXCLUSIVE PRICE
    646.00 €
    612.15 € / 5kg
    Save 33.85 €
    5% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 99-76-3
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
InChI Key LXCFILQKKLGQFO-UHFFFAOYSA-N
Synonym methylparaben, methyl paraben, methyl p-hydroxybenzoate, nipagin, methyl parahydroxybenzoate, maseptol, p-hydroxybenzoic acid methyl ester, 4-hydroxybenzoic acid methyl ester, p-carbomethoxyphenol, tegosept m
PubChem CID 7456
ChEBI CHEBI:31835
IUPAC Name methyl 4-hydroxybenzoate
SMILES COC(=O)C1=CC=C(C=C1)O

Specifications

Color White
Quantity 5 kg
Formula Weight 152.1
Percent Purity 99.83%
Physical Form Crystalline Powder
Chemical Name or Material Methyl-p-hydroxybenzoate
Product Identifier
  • Methyl-p-Hydroxybenzoate
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Methyl p-Hydroxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.