missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-tolyl sulfide, 97%

Product Code. 11317905
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317905 25 g 25g
11307905 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317905 Supplier Thermo Scientific Alfa Aesar Supplier No. L02933.14
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 623-13-2 | C8H10S | 138.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-13-2
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.23
MDL Number MFCD00008560
InChI Key VHILIAIEEYLJNA-UHFFFAOYSA-N
Synonym methyl p-tolyl sulfide, 4-methylthioanisole, 4-methylthio toluene, p-methylthio toluene, benzene, 1-methyl-4-methylthio, p-tolyl methyl sulfide, p-methylphenyl methyl sulfide, methyl p-cresyl sulfide, p-cresyl methyl sulfide
PubChem CID 69334
IUPAC Name 1-methyl-4-methylsulfanylbenzene
SMILES CSC1=CC=C(C)C=C1

Specifications

Density 1.029
Boiling Point 81°C to 82°C (18 mmHg)
Flash Point 85°C (185°F)
Odor Unpleasant
Refractive Index 1.573
Quantity 25 g
UN Number UN3334
Beilstein 1905733
Formula Weight 138.23
Percent Purity 97%
Chemical Name or Material Methyl p-tolyl sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.