missing translation for 'onlineSavingsMsg'
Learn More

Methyl Paraben, MedChemExpress

Product Code. 16499056
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499056 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16499056 Supplier Medchem Express Supplier No. HYN0349100mg
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Methyl Paraben, isolated from the barks of Tsuga dumosa the methyl ester of p-hydroxybenzoic acid, is a standardized chemical allergen. Methyl Paraben is a stable, non-volatile compound used as an antimicrobial preservative in foods, drugs and cosmetics. The physiologic effect of Methyl Paraben is by means of increased histamine release, and cell-mediated immunity.

Methyl Paraben, isolated from the barks of Tsuga dumosa the methyl ester of p-hydroxybenzoic acid, is a standardized chemical allergen. Methyl Paraben is a stable, non-volatile compound used as an antimicrobial preservative in foods, drugs and cosmetics. The physiologic effect of Methyl Paraben is by means of increased histamine release, and cell-mediated immunity.

Specifications

Chemical Name or Material Methyl Paraben
CAS 99-76-3
Color White to Off-White
Purity Grade Notes Research
Molecular Formula C8H8O3
Quantity 100 mg
Synonym Methyl 4-hydroxybenzoate
Solubility Information DMSO : 100 mg/mL (657.25 mM; Need ultrasonic)
SMILES O=C(OC)C1=CC=C(O)C=C1
Molecular Weight (g/mol) 152.15
Formula Weight 152.15
Percent Purity 99.56%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.