missing translation for 'onlineSavingsMsg'
Learn More

Methyl pentafluorobenzoate, 99%

Product Code. 15539343 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15539343

Brand: Thermo Scientific Acros 312260250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 36629-42-2 | C8H3F5O2 | 226.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36629-42-2
Molecular Formula C8H3F5O2
Molecular Weight (g/mol) 226.10
MDL Number MFCD00012172
InChI Key UXJRQNXHCZKHRJ-UHFFFAOYSA-N
Synonym methyl pentafluorobenzoate, pentafluorobenzoic acid methyl ester, benzoic acid, pentafluoro-, methyl ester, benzoic acid,2,3,4,5,6-pentafluoro-, methyl ester, methylpentafluorobenzoate, methyl perfluorobenzoate, acmc-209inq, methyl pentafluorbenzoate, perfluorobenzoic acid methyl ester
PubChem CID 606853
IUPAC Name methyl 2,3,4,5,6-pentafluorobenzoate
SMILES COC(=O)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Density 1.5300g/mL
Boiling Point 87.0°C to 88.0°C (20.0 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula F5C6CO2CH3
Refractive Index 1.4300 to 1.4320
Quantity 25 g
Specific Gravity 1.53
Formula Weight 226.1
Percent Purity 99%
Chemical Name or Material Methyl pentafluorobenzoate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.