missing translation for 'onlineSavingsMsg'
Learn More

Methyl pentafluorophenyl carbonate, 97%, Thermo Scientific Chemicals

Product Code. 11312897
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312897 25 g 25g
11302897 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312897 Supplier Thermo Scientific Alfa Aesar Supplier No. L16107.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

Methyl pentafluorophenyl carbonate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl pentafluorophenyl carbonate

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36919-03-6
Molecular Formula C8H3F5O3
Molecular Weight (g/mol) 242.10
MDL Number MFCD01075723
InChI Key HGYOVHMDBHQLOE-UHFFFAOYSA-N
Synonym methyl pentafluorophenyl carbonate, carbonic acid, methyl pentafluorophenyl ester, acmc-20akf4, pentafluorophenoxy methoxy ketone, methyl perfluorophenyl carbonate, carbonic acid methyl pentafluorophenyl ester
PubChem CID 14189360
SMILES COC(=O)OC1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 27°C to 31°C
Boiling Point 84°C (3 mmHg)
Quantity 25 g
UN Number UN2811
Beilstein 2289710
Solubility Information Insoluble in water.
IUPAC Name methyl 2,3,4,5,6-pentafluorophenyl carbonate
Formula Weight 242.1
Percent Purity 97%
Chemical Name or Material Methyl pentafluorophenyl carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.