missing translation for 'onlineSavingsMsg'
Learn More

Methyl phenyl carbonate, 97%

Product Code. 11400758
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400758 1 g 1g
11410758 5 g 5g
11420758 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400758 Supplier Thermo Scientific Alfa Aesar Supplier No. B25232.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 13509-27-8 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13509-27-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD04039782
InChI Key XTBFPVLHGVYOQH-UHFFFAOYSA-N
Synonym carbonic acid, methyl phenyl ester, phenyl methyl carbonate, phenyl methoxyformate, acmc-20akf2, carbonic acid methyl phenyl
PubChem CID 139482
IUPAC Name methyl phenyl carbonate
SMILES COC(=O)OC1=CC=CC=C1

Specifications

Boiling Point 210°C to 213°C
Refractive Index 1.521
Quantity 1 g
Formula Weight 152.15
Percent Purity 97%
Chemical Name or Material Methyl phenyl carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.