missing translation for 'onlineSavingsMsg'
Learn More

Methyl phenyl sulfoxide, 98+%

Product Code. 11437247
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11437247 100 g 100g
11417247 5 g 5g
11427247 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11437247 Supplier Thermo Scientific Alfa Aesar Supplier No. A15009.22
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1193-82-4 | C7H8OS | 140.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-82-4
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00002088
InChI Key JXTGICXCHWMCPM-UHFFFAOYSA-N
Synonym methyl phenyl sulfoxide, methylsulfinyl benzene, thioanisole s-oxide, phenyl methyl sulfoxide, sulfoxide, methyl phenyl, benzene, methylsulfinyl, methanesulfinylbenzene, methylphenylsulfoxide, methylsulphinyl benzene
PubChem CID 14516
IUPAC Name methylsulfinylbenzene
SMILES CS(=O)C1=CC=CC=C1

Specifications

Melting Point 26°C to 32°C
Boiling Point 93°C to 94°C (0.3 mmHg)
Flash Point 86°C (186°F)
Refractive Index 1.5775
Quantity 100 g
Beilstein 1904976
Sensitivity Hygroscopic
Formula Weight 140.2
Percent Purity ≥98%
Chemical Name or Material Methyl phenyl sulfoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.