missing translation for 'onlineSavingsMsg'
Learn More

Methyl salicylate, 99%

Product Code. 10032970 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
500mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10032970 1 L Glass bottle 1 L
10194700 5 mL Glass bottle 5mL
10606052 500 mL Glass bottle 500mL
10687744 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10032970 Supplier Thermo Scientific Acros Supplier No. 220490010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 119-36-8 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-36-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002214
InChI Key OSWPMRLSEDHDFF-UHFFFAOYSA-N
Synonym methyl salicylate, wintergreen oil, gaultheria oil, betula oil, teaberry oil, sweet birch oil, oil of wintergreen, analgit, spicewood oil, 2-hydroxybenzoic acid methyl ester
PubChem CID 4133
ChEBI CHEBI:31832
IUPAC Name methyl 2-hydroxybenzoate
SMILES COC(=O)C1=CC=CC=C1O

Specifications

Melting Point -8°C to 7°C
Color Colorless to Yellow
Density 1.1825g/mL
Boiling Point 222.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula 2-(HO)C6H4CO2CH3
Refractive Index 1.5350 to 1.5380
Quantity 1 L
Beilstein 10,7
Merck Index 15,6193
Specific Gravity 1.1825
Solubility Information Solubility in water: 0.07 % (20°C). Other solubilities: soluble in glacial acetic acid, soluble in ether and chloroform, in ethanol 80 vol. %:1:2 v/v (20°C), diethyl phthalate:1:1 v/v (20°C), 1,2-propanediol:1/7 v/v (20°C)
Formula Weight 152.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl salicylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.