Product Code. 12979284
Shop All SPEX Certiprep Products
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
unitSize
12979284
1 mL
1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code.
12979284
Supplier
SPEX Certiprep
Supplier No.
S2454
Please sign in to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued. View the product page for possible alternatives.
View alternative products
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte CAS No. Concentration Methyl Sulfide 75-18-3 1000μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Packaging
2mL Amber Prescored Glass Ampule
CAS
75-18-3, 67-56-1
CAS Min %
0.1, 99.9
Concentration or Composition Notes
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max %
0.1, 99.9
Molecular Formula
C2H6S
MDL Number
MFCD00008562
For Use With (Equipment)
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key
QMMFVYPAHWMCMS-UHFFFAOYSA-N
SMILES
CSC
IUPAC Name
(methylsulfanyl)methane
Solvent or Matrix
Methanol-P&T
Molecular Weight (g/mol)
62.13
PubChem CID
1068
Quantity
1 mL
ChEBI
CHEBI:17437
Chemical Name or Material
Methyl Sulfide
Show More
Show Less
Product intended for laboratory use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?