missing translation for 'onlineSavingsMsg'
Learn More

Methyl thiosalicylate, 97%

Product Code. 11442030
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11442030 5 g 5g
11452030 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11442030 Supplier Thermo Scientific Alfa Aesar Supplier No. H28400.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 4892-02-8 | C8H8O2S | 168.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4892-02-8
MDL Number MFCD00060678

Specifications

Density 1.223
Boiling Point 98°C to 100°C (2 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C8H7ClO2
Refractive Index 1.592
Quantity 5 g
Sensitivity Air Sensitive
Synonym 3-chloroanisaldehyde, 3-chloro-p-anisaldehyde, benzaldehyde, 3-chloro-4-methoxy, 3-chloro-4-methoxybenzenecarbaldehyde, 3-chloro-4-methoxy-benzaldehyde, pubchem2638, acmc-1ap2w, 3-chloro-4-methoxybenzaldehyde
InChI Key WYVGYYIZXPXHAZ-UHFFFAOYSA-N
SMILES COC1=C(C=C(C=C1)C=O)Cl
IUPAC Name 3-chloro-4-methoxybenzaldehyde
Molecular Weight (g/mol) 170.59
PubChem CID 78619
Formula Weight 168.21
Percent Purity 97%
Chemical Name or Material Methyl thiosalicylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.