missing translation for 'onlineSavingsMsg'
Learn More

Methyl Triphenylmethyl Ether, TRC

Product Code. 30482859
Change view
Click to view available options
Quantity:
50 mg
100 mg
5 g
Unit Size:
100mg
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482859 5 g 5g
30479490 100 mg 100mg
30477648 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482859 Supplier Toronto Research Chemicals Supplier No. TRCM3309755G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 596-31-6
Molecular Formula C20 H18 O
Molecular Weight (g/mol) 274.36
Synonym 1,1',1''-(Methoxymethylidyne)tris-benzene, Ether, methyl trityl (6CI,7CI,8CI), 1,1',1''-(Methoxymethylidyne)tris[benzene], Methoxytriphenylmethane, Methyl triphenylmethyl ether, Methyl trityl ether, NSC 64, Triphenylmethoxymethane, Triphenylmethyl methyl ether, Trityl methyl ether
IUPAC Name [methoxy(diphenyl)methyl]benzene
SMILES COC(c1ccccc1)(c2ccccc2)c3ccccc3

Specifications

Quantity 5 g
InChI Formula InChI=1S/C20H18O/c1-21-20(17-11-5-2-6-12-17,18-13-7-3-8-14-18)19-15-9-4-10-16-19/h2-16H,1H3
Formula Weight 274.1358
Chemical Name or Material Methyl Triphenylmethyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.