missing translation for 'onlineSavingsMsg'
Learn More

Methyl vanillate, 99%

Product Code. 11411257
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411257 5 g 5g
11421257 25 g 25g
11431257 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11411257 Supplier Thermo Scientific Alfa Aesar Supplier No. B24209.06
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 3943-74-6 | C9H10O4 | 182.175 g/mol

Methyl vanillate is used to investigate the volatile compounds of heather honeys from Calluna vulgaris and Erica arborea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl vanillate is used to investigate the volatile compounds of heather honeys from Calluna vulgaris and Erica arborea.

Solubility
Soluble in chloroform and methanol.

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 3943-74-6
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00008438
InChI Key BVWTXUYLKBHMOX-UHFFFAOYSA-N
Synonym methyl vanillate, methyl 3-methoxy-4-hydroxybenzoate, vanillic acid, methyl ester, 4-hydroxy-3-methoxybenzoic acid methyl ester, methylvanillate, benzoic acid, 4-hydroxy-3-methoxy-, methyl ester, vanillic acid methyl ester, unii-2hxg8qso3d, 2hxg8qso3d, vxx
PubChem CID 19844
ChEBI CHEBI:46477
IUPAC Name methyl 4-hydroxy-3-methoxybenzoate
SMILES COC1=C(C=CC(=C1)C(=O)OC)O

Spezifikation

Melting Point 62°C to 66°C
Boiling Point 285°C to 287°C
Quantity 5 g
Beilstein 1369113
Solubility Information Soluble in chloroform and methanol.
Formula Weight 182.18
Percent Purity 99%
Chemical Name or Material Methyl vanillate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.