missing translation for 'onlineSavingsMsg'
Learn More

Methyl vanillate, 99%

Product Code. 10136130 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10136130 5 g Glass bottle 5g
10002050 25 g Glass bottle 25g
10310214 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10136130 Supplier Thermo Scientific Acros Supplier No. 160300050
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 3943-74-6 | C9H10O4 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3943-74-6
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00008438
InChI Key BVWTXUYLKBHMOX-UHFFFAOYSA-N
Synonym methyl vanillate, methyl 3-methoxy-4-hydroxybenzoate, vanillic acid, methyl ester, 4-hydroxy-3-methoxybenzoic acid methyl ester, methylvanillate, benzoic acid, 4-hydroxy-3-methoxy-, methyl ester, vanillic acid methyl ester, unii-2hxg8qso3d, 2hxg8qso3d, vxx
PubChem CID 19844
ChEBI CHEBI:46477
IUPAC Name methyl 4-hydroxy-3-methoxybenzoate
SMILES COC1=C(C=CC(=C1)C(=O)OC)O

Specifications

Melting Point 64.0°C to 67.0°C
Color Beige to Yellow
Boiling Point 285.0°C to 287.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HOC6H3(OCH3)CO2CH3
Quantity 5 g
Beilstein 10, 396
Formula Weight 182.18
Percent Purity 99%
Physical Form Powder or Granules
Chemical Name or Material Methyl vanillate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl vanillate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.