missing translation for 'onlineSavingsMsg'
Learn More

Methyl vanillate, MedChemExpress

Product Code. 16421969
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16421969 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16421969 Supplier Medchem Express Supplier No. HY75342
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Methyl vanillate, one of the ingredients in Hovenia dulcis Thunb, is a Wnt/β-catenin pathway activator. A benzoate ester that is the methyl ester of vanillic acid. It has a role as an antioxidant and a plant metabolite.

Methyl vanillate, one of the ingredients in Hovenia dulcis Thunb, is a Wnt/β-catenin pathway activator. A benzoate ester that is the methyl ester of vanillic acid. It has a role as an antioxidant and a plant metabolite.

Specifications

Chemical Name or Material Methyl vanillate
CAS 3943-74-6
Color White
Purity Grade Notes Research
Molecular Formula C9H10O4
Quantity 1 g
Solubility Information DMSO : ≥ 100 mg/mL (548.94 mM) ∣H2O : 1.67 mg/mL (9.17 mM; Need ultrasonic)
SMILES O=C(OC)C(C=C1)=CC(OC)=C1O
Molecular Weight (g/mol) 182.17
Formula Weight 182.17
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.