missing translation for 'onlineSavingsMsg'
Learn More

Methylamine hydrochloride, 99%

Product Code. 10488880 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
500 g
1 kg
5 kg
Unit Size:
1kg
250g
500g
50g
5kg
This item is not returnable. View return policy

Product Code. 10488880

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 593-51-1 | CH6ClN | 67.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-51-1
Molecular Formula CH6ClN
Molecular Weight (g/mol) 67.52
InChI Key NQMRYBIKMRVZLB-UHFFFAOYSA-N
IUPAC Name methanaminium chloride
SMILES [Cl-].C[NH3+]

Specifications

Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula CH3NH2·HCl
Quantity 250 g
Formula Weight 67.52
Chemical Name or Material Methylamine hydrochloride
compliance-icons
Product Identifier
  • Methylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Methanamine, hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.