missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyle2-amino-3-(1H-indol-3-yl)-2-methylpropanoate, 97%, Maybridge™
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| CAS Min % | 97.0 |
| Assay Percent Range | 97% |
| Packaging | Amber glass bottle |
| Quantity | 1 g |
| Formula Weight | 232.28 |
| Percent Purity | 97% |
| Chemical Name or Material | Methyle2-amino-3-(1H-indol-3-yl)-2-methylpropanoate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction