missing translation for 'onlineSavingsMsg'
Learn More

Methyle2-chloro-3,4-dimethoxybenzoate, 97%, Thermo Scientific™

Product Code. 10744763
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10744763 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10744763 Supplier Thermo Scientific Maybridge Supplier No. BTB09269EA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 175136-01-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H11ClO4
MDL Number MFCD00085114
Quantity 10 g
Synonym maybridge1_001981, benzoic acid,2-chloro-3,4-dimethoxy-, methyl ester
InChI Key RYYAHUFRPCBHCT-UHFFFAOYSA-N
SMILES COC1=C(C(=C(C=C1)C(=O)OC)Cl)OC
IUPAC Name methyl 2-chloro-3,4-dimethoxybenzoate
Molecular Weight (g/mol) 230.644
PubChem CID 2774981
Formula Weight 230.65
Percent Purity 97%
Chemical Name or Material Methyle2-chloro-3, 4-dimethoxybenzoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.